Kimax Media Bottles
Mix, store, and transport culture media, chemicals, and solvents
These heavy-duty general-purpose Type 1 borosilicate glass bottles have permanent white enamel graduations and marking areas. Each bottle includes an autoclavable (140°C/284°F) linerless blue polypropylene (PP) cap and drip-free pour ring. Maximum temperature: 450°C (840°F). Autoclavable.
4,300.00 - ₹42,500.00INR / Case of 10
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
DWK Life Sciences (Kimble) Media Bottle ISO, GL45, 100 ml, Blue Polypropylene Cap 10/Cs UL-15957-04 Mfr # SP14395-100
COMPARE
₹6,400.00INR / Case of 10
DWK Life Sciences (Kimble) Media Bottle ISO, GL45, 250 ml, Blue Polypropylene Cap 10/Cs UL-15957-05 Mfr # SP14395-250
COMPARE
₹7,100.00INR / Case of 10
DWK Life Sciences (Kimble) Media Bottle ISO, GL45, 500 ml, Blue Polypropylene Cap 10/Cs UL-15957-06 Mfr # SP14395-500
COMPARE
₹8,800.00INR / Case of 10
DWK Life Sciences (Kimble) Media Bottle ISO, GL45, 1000 ml, Blue Polypropylene Cap 10/Cs UL-15957-07 Mfr # SP14395-1000
COMPARE
₹14,200.00INR / Case of 10
DWK Life Sciences (Kimble) Media Bottle ISO, GL45, 2000 ml, Blue Polypropylene Cap/10Cs UL-15957-08 Mfr # SP14395-2000
COMPARE
₹31,500.00INR / Case of 10
DWK Life Sciences (Kimble) Bottle Media, 5 L, 1/Cs UL-15957-09 Mfr # SP14395-5000
COMPARE
₹20,200.00INR / Case of 1
DWK Life Sciences (Kimble) Bottle Media, 10 L, 1/Cs UL-15957-10 Mfr # SP14395-10000
COMPARE
₹37,300.00INR / Case of 1
DWK Life Sciences (Kimble) Bottle Media, 20 L, 1/Cs UL-15961-72 Mfr # SP14395-20000
COMPARE
₹42,500.00INR / Case of 1
DWK Life Sciences (Kimble) Blue Polypropylene Cap, 10/Cs UL-15961-73 Mfr # 14395C-45
COMPARE
₹4,300.00INR / Case of 10
Showing 1- 9 of 9