KIMAX Glass Filtering Flasks
These graduated flasks have a heavier wall than standard Erlenmeyer flasks, and can withstand 1 atmosphere, or vacuum to 29" Hg. Tubulation OD for flasks from 25- to 500-mL capacity is 5⁄16". Tubulation OD for 1000- to 4000-mL flasks is 3⁄8". Compliance: ASTM E 1406, Type II, Class I. Temperature range: 840°F (450°C) max.
45,300.00 - ₹328,000.00INR / Case of 18
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Stopper Number Accepted
Availability Pricing
DWK Life Sciences (Kimble) Kimax Filtering Glass Flask, 25 mL, 5/16" barb, 18/cs WW-34524-30 Mfr # 27060-25
COMPARE
25 Glass 3
₹181,000.00INR / Case of 18
DWK Life Sciences (Kimble) Kimax Filtering Glass Flask, 50 mL, 5/16" barb, 18/cs WW-34524-32 Mfr # 27060-50
COMPARE
50 Glass 4
₹209,000.00INR / Case of 18
DWK Life Sciences (Kimble) Kimax Filtering Glass Flask, 125 mL, 5/16" barb, 18/cs WW-34524-34 Mfr # 27060-125
COMPARE
125 Glass 5
₹225,000.00INR / Case of 18
DWK Life Sciences (Kimble) Kimax Filtering Glass Flask, 500 mL, 5/16" barb, 18/cs WW-34524-38 Mfr # 27060-500
COMPARE
500 Glass 7
₹241,000.00INR / Case of 18
DWK Life Sciences (Kimble) Kimax Filtering Glass Flask, 1000 mL, 3/8" barb, 12/cs WW-34524-40 Mfr # 27060-1000
COMPARE
1000 Glass 8
₹328,000.00INR / Case of 12
DWK Life Sciences (Kimble) Kimax Filtering Glass Flask, 2000 mL, 3/8" barb, 1/cs WW-34524-42 Mfr # 27060-2000
COMPARE
2000 Glass 9
₹45,300.00INR / Case of 1
DWK Life Sciences (Kimble) Kimax Filtering Glass Flask, 4000 mL, 3/8" barb, 1/cs WW-34524-44 Mfr # 27060-4000
COMPARE
4000 Glass 12
₹68,500.00INR / Case of 1
Showing 1- 7 of 7