Kimax Class B Graduated Cylinders
Calibrated “to deliver” these single metric scale graduated cylinders meet ASTM Specification E1272, Style 1, Class B requirements
  • Single metric scale
Bumpers are included with 25- to 2000-mL cylinders. Hexagonal base on all cylinders increases stability; the 10- and 25-mL cylinders are comparatively shorter for even greater stability. The enlarged funnel-shaped top on the 10-mL cylinder makes filling easy.
104,000.00 - ₹170,000.00INR / Case of 24
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
DWK Life Sciences (Kimble) 20024-25 Graduated Glass Cylinders, to deliver, 25 mL WW-34527-02 Mfr # 20024-25
COMPARE
25 0.5 Glass
₹120,000.00INR / Case of 24
DWK Life Sciences (Kimble) 20024-100 100 mL class B graduated cylinders WW-34527-06 Mfr # 20024-100
COMPARE
100 1 Glass
₹170,000.00INR / Case of 24
DWK Life Sciences (Kimble) 20024-250 250 mL class B graduated cylinders WW-34527-08 Mfr # 20024-250
COMPARE
250 2 Glass
₹117,000.00INR / Case of 12
DWK Life Sciences (Kimble) 20024-500 500 mL class B graduated cylinders WW-34527-10 Mfr # 20024-500
COMPARE
500 5 Glass
₹104,000.00INR / Case of 8