Kapton Polymide Tape
Polymide tape is a chemical-resistant tape that is manufactured with a high temperature silicone adhesive. Provides high heat resistance and tensile strength, good solvent resistance, and excellent dielectric strength. Temperature range is -100 to 500°F (-73 to 260°C). Meets UL-510 for flame retardant specifications.
3,100.00 - ₹15,500.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material Thickness (mil)
Availability Pricing
COMPARE
36 Polymide 2.5
₹3,100.00INR / Each
COMPARE
36 Polymide 2.5
₹4,300.00INR / Each
COMPARE
36 Polymide 2.5
₹9,000.00INR / Each
COMPARE
36 Polymide 2.5
₹7,800.00INR / Each
COMPARE
36 Polymide 2.5
₹10,400.00INR / Each
COMPARE
36 Polymide 2.5
₹15,500.00INR / Each