Kanomax Handheld Micromanometer and Velocity/Flow Kits
View and analyze data on the go
  • USB and Bluetooth® communicate with PCs and Android™-based smart phones or tablets
  • Ideal for HVAC, TAB, and Cleanroom certification applications
The manometer transmits data from its expansive memory to PC or tablet via included software or the free mobile app downloadable from the manufacturer's website for immediate analysis.
Choose the model 6700 manometer by itself (10007-00) for use with existing pitot tubes, velocity grids, and flow grids. Or choose the velocity grid kit (10007-01) or air capture hood kit (10007-11) — both include everything you need to begin monitoring air velocity and/or flow.
Note: The Bluetooth® word mark and logos are registered trademarks owned by Bluetooth SIG, Inc.
686,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kanomax 6700 Handheld Micromanometer with Velocity Grid WW-10007-01 Mfr # 6700-VG
COMPARE
Handheld Micromanometer with Velocity Grid
₹686,000.00INR / Each