Justrite Sure-Grip® EX Undercounter Flammable Safety Cabinets
Save space and safely store your flammable liquids in sturdy undercounter cabinets
  • FM approved
  • OSHA and NFPA Compliant
Undercounter cabinets are constructed of double-walled, 18-gauge steel. Recessed toe kick improves ergonomics by allowing you to stand closer to work surface. Powder-coated cabinets feature dual back wall vents with flame arrester to keep contents cool and prevent build-up of dangerous fumes. Includes a 3-point bullet latch for failsafe closure and a SpillSlope® shelf that directs spills to back and down to sump. Adjust shelf in 2¼" increments to maximize storage space.
248,000.00 - ₹312,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Self-Closing
Availability Pricing
Justrite 892320 Sure-Grip® EX Undercounter Flammable Safety Cabinet, Self Close, 22 Gal WW-81790-24 Mfr # 892320
COMPARE
22 Yes
₹312,000.00INR / Each
Justrite 892300 Sure-Grip® EX Undercounter Flammable Safety Cabinet, Manual Close, 22 Gal WW-81790-25 Mfr # 892300
COMPARE
22 No
₹248,000.00INR / Each