Justrite Steel Safety Drum Funnels
Prevent spills when transferring flammable solvents
  • Hinged cover reduces spread of potentially flammable vapors
  • 2" NPS connector
Drum funnels feature a wide-mouth design for safe and fast pouring of liquids without spilling. All funnels are constructed with a galvanized steel, durable powder-coat finish and feature a 2" NPS connector and a hinged cover to reduce the spread of potentially flammable vapors.
24,400.00 - ₹65,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in) Top OD (in)
Availability Pricing
Justrite 08202 Steel Safety Drum Funnel, Manual Closing Cover, 1" Flame Arrester, 2" Bung WW-09934-32 Mfr # 08202
COMPARE
Steel 4 1/2 4 1/2
₹24,400.00INR / Each
Justrite 08207 Steel Safety Drum Funnel, Self-Closing Cover, 6" Brass Flame Arrester, 2" Bung WW-09934-34 Mfr # 08207
COMPARE
Steel 10 10 3/4
₹65,100.00INR / Each