Justrite Sure-Grip® EX Slimline Flammable Safety Cabinets
Extra depth for maximum capacity in a slim-width cabinet
  • FM approved
Safely store flammables in these cabinet when floor space is tight or use to expand storage alongside existing cabinets. The cabinets are fire tested, FM approved, and complies with OSHA and NFPA. Safety features include sturdy 18-gauge (1 mm) thick double-wall, welded steel with 1 ½” (38 mm) of insulating air space, and a 3-point stainless steel bullet latching system. Adjustable, patented SpillSlope® galvanized steel shelves direct spills to 2” (51 mm) leakproof sump.
Features include U-Loc™ handle with two keys (accepts optional padlock), trilingual warning with Haz-Alert™ reflective labels, dual vents with flame arresters, adjustable leveling feet, and grounding connector.
Self-closing models equipped with fusible link holds door open during use and melts at 165°F (74° C) to automatically close under fire conditions.
134,000.00 - ₹323,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Self-Closing
Availability Pricing
Justrite 892220 Sure-Grip® EX Slimline Flammable Safety Cabinet, Self Close, 1 Door, 22 Gal WW-86526-06 Mfr # 892220
COMPARE
22 Yes
₹323,000.00INR / Each
Justrite 892200 Sure-Grip® EX Slimline Flammable Safety Cabinet, Manual Close, 1 Door, 22 Gal WW-86526-08 Mfr # 892220
COMPARE
22 No
₹134,000.00INR / Each
Justrite 895420 Sure-Grip® EX Deep Slimline Flammable Safety Cabinet, Self Close, 1 Door, 54 Gal WW-86526-10 Mfr # 895420
COMPARE
54 Yes
₹299,000.00INR / Each