This product is now discontinued, View Related Products
Julabo HighTech Heating Circulating Baths, Single Program
Highly stable temperature control improves system performance
  • Ensure repeatable results with ±0.01°C stability
  • Durable stainless-steel tanks withstand higher temperatures
  • RS-232 interface for optional remote control
  • Integrated cooling coil connects with tap water supply
  • Ideal for both internal and external circulation applications
These heating circulators offer superior temperature stability on all models and RS-232 interface for remote control. External pump connections and integrated cooling coils enhance the versatility of these circulating baths.
HighTech (HE, SE series) circulators feature four-stage force and suction pumps for external closed loop and open system applications. The HE and SE series circulators offer basic multistep programming and data monitoring capabilities.
919,000.00 - ₹999,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Reservoir Capacity (L) Temperature Stability (° C)
Availability Pricing
Julabo HE-4 HighTech Programmable Heating Circulator, Single Program, 4.5L; 100 to 115 VAC, 50/60 Hz WW-12151-02 Mfr # 9212504.22
COMPARE
100 to 115 4.5 ±0.01
Call For Price
Julabo HE-4 HighTech Programmable Heating Circulator, Single Program, 4.5L; 208 to 230 VAC, 50/60 Hz WW-12151-03 Mfr # 9212504.33
COMPARE
208 to 230 4.5 ±0.01
Call For Price
Julabo SE-6 HighTech Programmable Heating Circulator, Single Program, 6L; 208 to 230 VAC, 50/60 Hz WW-12151-04 Mfr # 9252506.33
COMPARE
208 to 230 6 ±0.01
₹919,000.00INR / Each
Julabo SE-12 HighTech Programmable Heating Circulator, Single Program, 12L; 208 to 230 VAC, 50/60 Hz WW-12151-05 Mfr # 9252512.33
COMPARE
208 to 230 12 ±0.01
₹924,000.00INR / Each
Julabo SE-26 HighTech Programmable Heating Circulator, Single Program, 26L; 208 to 230 VAC, 50/60 Hz WW-12151-06 Mfr # 9252526.33
COMPARE
208 to 230 26 ±0.01
₹999,000.00INR / Each