This product is now discontinued, View Related Products
Julabo FD Series Flow-Through Chiller
Rapidly cools fluid down below ambient temperature
  • An economical alternative to conventional tap water cooling
  • Compact design maximizes bench space
  • Built-in handle makes moving unit around the lab easy
  • Easy to operate and maintain
  • Low energy requirement
The Julabo FD series flow-through chiller can easily be connected to open bath circulators, immersion circulators, and bridged-mounted circulators to help extend their temperature range to below ambient temperature. The chiller is ideal for a variety of cooling applications and offers an economical solution to conventional tap water cooling. Integrates into an existing cooling loop circuit of a closed system. Features a flow rate of 2 to 3 L/min.
1,084,000.00 - ₹1,171,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 20° C (watts) Min Temperature (° C) Power (VAC)
Availability Pricing
Julabo FD200 Flow-Through Chiller, 115V/60Hz WW-12150-49 Mfr # 9655825.2
COMPARE
220 10 115
₹1,084,000.00INR / Each
Julabo FD200 Flow-Through Chiller, 230V/50Hz WW-12150-50 Mfr # 9655825.3
COMPARE
220 10 230
₹1,171,000.00INR / Each