This product is now discontinued, View Related Products
Julabo Calibration Baths
Deliver excellent temperature stability and uniformity
  • Intelligent Cascade Control offers self-optimizing adjustment of PID control parameters
  • Temperature control features allow access to all temperature control parameters
  • Early warning system alerts you when liquid level is low
  • Adjustable high temperature shutoff protects bath and application from adverse events
  • Suitable for use with flammable bath fluids—Class III (FL)
Ideally suited for applications in calibration laboratories, these calibration baths offer a temperature stability of ±0.005°C and a temperature uniformity of ±0.01°C. The Absolute Temperature Calibration (ATC3) provides high temperature stability in the bath and offers three-point calibration. Smart pump technology offers consistent pump performance with the electronically adjustable pump stages. An integrated programmer stores and runs up to 360 different temperature and time-dependent processes.
An easy-to-read VFD displays warning functions, high temperature cutoff, and pump stages. The backlit LCD displays interactive operation in plain text, while the splashproof keypad lets you easily access menu functions. A heated cover plate prevents condensation or ice build-up in the bath. Other features include a Pt100 external sensor connection, automatic operating time control, built-in RS-232/RS485 interface, and a removable venting grid for quick and easy cleaning.
2,941,000.00 - ₹4,619,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Min Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
Julabo FK30-SL Calibration Bath, 14 L; 115V/60Hz WW-12150-88 Mfr # 9352627.02
COMPARE
-22 115 14
₹3,859,000.00INR / Each
Julabo FK30-SL Calibration Bath, 14 L; 230V/50Hz WW-12150-89 Mfr # 9352627.03
COMPARE
-22 230 14
₹4,168,000.00INR / Each
Julabo FK31-SL Calibration Bath, 24 L; 115V/60Hz WW-12150-90 Mfr # 9352628.02
COMPARE
-22 115 24
₹4,264,000.00INR / Each
Julabo FK31-SL Calibration Bath, 24 L; 230V/50Hz WW-12150-91 Mfr # 9352628.03
COMPARE
-22 230 24
₹4,619,000.00INR / Each
Julabo SL-8K Calibration Bath, 8 L; 230V/50-60Hz WW-12150-92 Mfr # 9352508.33
COMPARE
122 230 8
₹2,941,000.00INR / Each
Julabo SL-14K Calibration Bath, 14 L; 230V/50-60Hz WW-12150-93 Mfr # 9352514.33
COMPARE
122 230 14
₹3,340,000.00INR / Each