Cole-Parmer® Essentials Stainless Steel Apparatus Positioners/Lab Jacks
Durable heavy-duty lab lifts offer an optional ratchet tool for heavy lifting with little effort
  • Constructed of 300 series stainless steel
  • Oversize grip adjustment knobs
  • Can be autoclaved or chemically cleaned
Control the height of your heavy labware products with these scissor jacks that feature ultra stable lifting platforms with exceptional strength.
38,500.00 - ₹329,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Description
Availability Pricing
COMPARE
20 Lab Jack, 3\" x 3\" Stainless Steel Lab-Lift, 20lb (9kg) Limit
₹38,500.00INR / Each
COMPARE
30 Lab Jack, 4\" x 4\" Stainless Steel Lab-Lift, 30lb (13.6kg) Limit
₹46,600.00INR / Each
COMPARE
35 Lab Jack, 6\" x 6\" Stainless Steel Lab-Lift, 35Lb (16kg) Limit
₹88,000.00INR / Each
COMPARE
55 Lab Jack, 8\" x 8\" Stainless Steel Lab-Lift, 55Lb (25kg) Limit
₹67,300.00INR / Each
COMPARE
80 Lab Jack, 10\" x 10\" Stainless Steel Lab-Lift, 80lb (36.3kg) Limit
₹85,400.00INR / Each
COMPARE
100 Lab Jack, 12\" x 12\" Stainless Steel Lab-Lift, 100 lb (45.4 kg) Limit
₹185,000.00INR / Each
COMPARE
180 Lab Jack, 16\" x 16\" Stainless Steel Lab-Lift, 180lb (81kg) Limit
₹329,000.00INR / Each
Showing 1- 7 of 7