JFP-70 Five-Channel Digital Flame Photometer
Simple to operate with large 7” color touch graphic screen
  • Direct concentration readings in mmol/L, mg/100 mL, and g/mL
  • Accurately measure five elements simultaneously: K+, Na+, Li+, Ca+, and Ba+
  • An automatic calculation of correlation coefficient and pre-selection of flame sizes
  • A flameout protection device for safety
  • Direct printing devices; (To be equipped with a printer)
  • Use USB interface to connect with computer and with software to do data processing
New digital flame photometer with 7” color touch screen and 5-channel simultaneous operation. Data can be transfer to computer through USB or can also be fitted with printer (Optional).
The system comes complete with air compressor, air filtration valve, sprayer, and atomizing chamber.
455,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Jenway JFP-70P Digital Flame Photometer with Inbuilt Printer; 1/Ea
₹455,000.00INR / Each