Cole-Parmer® FF-200 Series Economical Flame Photometer
Automatic gas shutoff feature ensures operator safety
  • Unit comes with Na+, K+, Li+, Ca+, and Ba+ filters
  • Low temperature and single channel
Use this economical flame photometer for routine determinations of sodium and potassium in industrial and educational applications. The fine and coarse adjustment controls provide accurate measurements every time. For safety, an automatic gas shutoff mechanism activates if the flame becomes accidentally extinguished.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Aspiration Rate Power (VAC) Range
Availability Pricing
COMPARE
2 to 6 mL/min 120 0 to 199.9 ppm
Call For Price
COMPARE
2 to 6 mL/min 230 0 to 199.9 ppm