This product is now discontinued, View Related Products

Jenway Colorimeter

Easy to use — ideal for routine and educational use
  • Wide wavelength range lets you measure across the visible spectrum
  • Versatile — operates on AC power or 12 V battery
  • Concentration range: 0.1 to 1000 (0.1 to 1 resolution)
This general-purpose benchtop colorimeter features eight built-in gelatin filters for use in commonly encountered determinations for the following wavelengths: 430, 470, 490, 520, 540, 580, 600 and 710 nm. Results are displayed directly in transmittance, absorbance, or concentration units on a LCD.


Included sample holder accommodates 10 x 10 mm cuvettes of standard or semi-micro volumes or test tubes.

201,000.00 - ₹230,000.00INR / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Benchtop Colorimeter with Filters; 230 V
₹201,000.00INR / Each
COMPARE
Benchtop Colorimeter with Filters; 120 V
₹230,000.00INR / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat