Jenway® 6305 Benchtop UV/Visible Spectrophotometer
An easy-to-use instrument with a compact footprint
  • Wavelength range of 198 to 1000 nm
  • Absorbance, %T, and concentration modes
  • Recalls previous settings at power up
  • Analog and RS-232 outputs let you interface with printers and PCs
  • Multi-parameter display shows error messages, prompts, mode indication, and concentration units
This easy-to-use spectrophotometer is ideal for the busy technician or student. The unit covers a wide range of wavelength from 198 to 1000 nm and provides fast warm-up time, low drift, and high reliability.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Wavelength (nm) Min Wavelength (nm) Power (VAC)
Availability Pricing
COMPARE
1000 198 230
Call For Price
COMPARE
1000 198 115