This product is now discontinued, View Related Products
Isocratic HPLC Systems
Instruments for your performance requirements and budgetary constraints
Our HPLC systems provide the functionality necessary to achieve precise high-quality separations for analytical and preparative methods. Select from PEEK or stainless steel construction with all the features needed for consistent results and total component integration for ease of use. Pumps utilize a built-in pulse damper for smooth, low-pulse flows. Features include solvent delivery, gradient/isocratic control, 20 µL injection loop, UV-Vis detection, and data acquisition and processing.
Isocratic HPLC Systems with Variable Wavelength Detectors have a small footprint, yet provide precision for routine separation and analysis of compounds absorbing in the 254 nm range. Ideal for analytical and preparative applications not requiring gradient capability. All wettable parts are const
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Max Wavelength (nm) Power (VAC)
Availability Pricing
COMPARE
3000 800 115
Call For Price
COMPARE
3000 800 230
Call For Price
COMPARE
3000 254 115
Call For Price
COMPARE
3000 254 230
Call For Price
COMPARE
5000 800 115
Call For Price
COMPARE
5000 800 230
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
5000 800
Call For Price
COMPARE
5000 800
Call For Price
Showing 1- 10 of 10