Ismatec® Microbore Pump Tubing, Peroxide-Cured Silicone
Reduce the risk of particulate entrapment with smooth inner surface
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Demanding high-pressure pharmaceutical, biotechnology, aerospace, and food and beverage applications.
16,500.00 - ₹17,500.00INR / Pkg of 1
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm)
Availability Pricing
COMPARE
0.25
₹16,500.00INR / Pkg of 1
COMPARE
0.38
₹16,500.00INR / Pkg of 1
COMPARE
0.51
₹16,500.00INR / Pkg of 1
COMPARE
0.64
₹16,500.00INR / Pkg of 1
COMPARE
0.76
₹16,500.00INR / Pkg of 1
COMPARE
0.89
₹17,100.00INR / Pkg of 1
COMPARE
1.02
₹16,500.00INR / Pkg of 1
COMPARE
1.14
₹17,500.00INR / Pkg of 1
COMPARE
1.3
₹16,500.00INR / Pkg of 1
COMPARE
1.42
₹17,100.00INR / Pkg of 1
COMPARE
1.52
₹16,500.00INR / Pkg of 1
COMPARE
1.65
₹16,500.00INR / Pkg of 1
COMPARE
1.85
₹16,500.00INR / Pkg of 1
COMPARE
2.06
₹17,100.00INR / Pkg of 1
COMPARE
2.29
₹16,500.00INR / Pkg of 1
COMPARE
2.54
₹16,500.00INR / Pkg of 1
COMPARE
2.79
₹17,100.00INR / Pkg of 1
Showing 1- 17 of 17