Ismatec® Pump Tubing, GORE® STA-PURE® Series PFL Single Elements
Excellent choice for high pressure, low spallation peristaltic tubing applications
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Engineered for long life in peristaltic pump applications
  • Unique composite of expanded PTFE and platinum-cured perfluoroelastomer
Applications: Chemical, critical pharmaceutical, and biopharma processing applications
Characteristics: High resilience and strength, low spallation and gas permeability. Opaque.
Certifications: USP Class VI compliant, REACH and RoHS compliant, and manufactured in Class 7 (ISO 146441-1) cleanroom, under cGMP certified to ISO 13485 and 15378.
Temperature range: -20 to 80 °C
Sterilize: Autoclave, CIP/SIP
The Ismatec® GORE® STA-PURE® Series PFL pump tubing is designed to withstand pumping aggressive chemicals with a composite structure made of platinum-cured perfluoroelastomer and ePTFE. Long-term testing showed pumping common organic solvents (acetic acid, acetone, and ethanol) at 10 psig did not reduce the pumping life of the tubing compared with pumping deionized water at the same pressure. Single element and double-Y element versions available.
156,000.00 - ₹1,806,000.00INR / Pkg of 1
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (m) Tubing ID (mm) Wall Thickness (mm)
Availability Pricing
COMPARE
0.355 2.8 0.8
₹156,000.00INR / Pkg of 1
COMPARE
0.305 1.6 1.6
₹156,000.00INR / Pkg of 1
COMPARE
0.355 1.6 1.6
₹173,000.00INR / Pkg of 1
COMPARE
0.305 3.2 1.6
₹168,000.00INR / Pkg of 1
COMPARE
0.355 3.2 1.6
₹182,000.00INR / Pkg of 1
COMPARE
0.305 4.8 1.6
₹168,000.00INR / Pkg of 1
COMPARE
0.355 4.8 1.6
₹183,000.00INR / Pkg of 1
COMPARE
0.305 6.4 1.6
₹173,000.00INR / Pkg of 1
COMPARE
0.355 1.6 2.4
₹190,000.00INR / Pkg of 1
COMPARE
0.355 3.2 2.4
₹196,000.00INR / Pkg of 1
COMPARE
0.355 6.4 2.4
₹208,000.00INR / Pkg of 1
COMPARE
0.61 9.5 2.4
₹381,000.00INR / Pkg of 1
COMPARE
0.61 9.5 3.2
₹389,000.00INR / Pkg of 1
COMPARE
0.61 12.7 3.2
₹475,000.00INR / Pkg of 1
COMPARE
0.61 15.9 3.2
₹570,000.00INR / Pkg of 1
COMPARE
0.38 12 4
₹333,000.00INR / Pkg of 1
COMPARE
0.38 17 4
₹438,000.00INR / Pkg of 1
COMPARE
0.61 12.7 4.8
₹814,000.00INR / Pkg of 1
COMPARE
0.61 15.9 4.8
₹1,037,000.00INR / Pkg of 1
COMPARE
0.305 25.4 4.8
₹1,806,000.00INR / Pkg of 1
Showing 1 - 20 of 28