This product is now discontinued, View Related Products
Instek Single output DC Switching Power Supplies
Ideal for power-efficient bench-top or portable applications requiring high regulation
  • Dual measurement display
  • 0.01% high regulation
  • ±0.5% of rdg + 2 digits accuracy
  • Constant voltage and constant current operation
  • High efficiency
  • High power density
  • Over voltage protection protects loads from unexpected conditions
  • Remote output on/off control
  • 115 to 230 VAC, 50/60 Hz
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
GW Instek SPS-1230 DC Switching Single Output Switching Power Supply, 12V, 30A WW-20042-55 Mfr # SPS-1230
COMPARE
0 to 30 A 0 to 12 VDC ≤ 5 mVrms, ≤ 100 mVp-p
Call For Price
GW Instek SPS-1820 DC Switching Single Output Switching Power Supply, 18V, 20A WW-20042-56 Mfr # SPS-1820
COMPARE
0 to 20 A 0 to 18 VDC ≤ 5 mVrms, ≤ 100 mVp-p
Call For Price
GW Instek SPS-2415 DC Switching Single Output Switching Power Supply, 24V, 15A WW-20042-57 Mfr # SPS-2415
COMPARE
0 to 15 A 0 to 24 VDC ≤ 5 mVrms, ≤ 100 mVp-p
Call For Price
GW Instek SPS-3610 DC Switching Single Output Switching Power Supply, 36V, 10A WW-20042-58 Mfr # SPS-3610
COMPARE
0 to 10 A 0 to 36 VDC ≤ 5 mVrms, ≤ 100 mVp-p
Call For Price
GW Instek SPS-606 DC Switching Single Output Switching Power Supply, 60V, 6A WW-20042-59 Mfr # SPS-606
COMPARE
0 to 6 A 0 to 60 VDC ≤ 5 mVrms, ≤ 100 mVp-p