Instek PSB-1000 Series Multirange DC Power Supply
No need to study the user manual—get up and running faster!
  • Menu-driven interface is user-friendly with plain language prompts
  • Constant power output works for multirange voltage and current operation
  • Connect multiple units in series or parallel to increase output
  • Program output sequences to begin immediately upon power-up
This series of power supplies features USB and LAN connectivity, with GPIB optional (order communication card separately), for remote programming and test sequence triggering. All programming options are available via keypad and LCD as well.
Apply triggers via BNC or program output delays in addition to the optional automatic sequence start upon power-up. Front and rear output terminals provide convenience, while rear-panel serial connector increases versatility by allowing series and parallel operation of multiple units in master/slave mode. When connecting units for series or parallel operation, all units must be the same model. Up to four units connect in parallel and two units connect in series.
342,000.00 - ₹485,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek PSB-1400L Programmable Multirange DC Power Supply, 400 W WW-20026-17 Mfr # PSB-1400L
COMPARE
Programmable Multirange DC Power Supply, 400 W
₹342,000.00INR / Each
GW Instek PSB-1400M Programmable Multirange DC Power Supply, 400 W WW-20026-19 Mfr # PSB-1400M
COMPARE
Programmable Multirange DC Power Supply, 400 W
₹342,000.00INR / Each
GW Instek PSB-1800L Programmable Multirange DC Power Supply, 800 W WW-20026-21 Mfr # PSB-1800L
COMPARE
Programmable Multirange DC Power Supply, 800 W
₹485,000.00INR / Each
GW Instek PSB-1800M Programmable Multirange DC Power Supply, 800 W WW-20026-23 Mfr # PSB-1800M
COMPARE
Programmable Multirange DC Power Supply, 800 W
₹485,000.00INR / Each