Instek High-Current Output Benchtop DC Power Supplies
Feature constant voltage and current operation with overload protection
Wire two single-output models with the same range in a master/slave circuit. A rear panel switch lets you select either master or slave with connections for series or parallel hook-up. Connect in series to double the voltage output; connect in parallel to double the current output. Single-output models also feature a HI/LO range button for current output; the LO range provides a better resolution through a portion of the range.
Triple output model features two independent variable power supplies and one 5 VDC fixed output. Easily set up a master/slave circuit with this model. Connect the two independent power supplies in series or parallel simply by pressing a button.
188,000.00 - ₹211,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
GW Instek GPR-1820HD High-Current, Single-Output DC Power Supply, 0 to 18 VDC, 0 to 20 A WW-26856-92 Mfr # GPR-1820HD
COMPARE
0 to 20 A 0 to 18 VDC ≤ 0.5 mVrms; ≤ 3 mArms
₹211,000.00INR / Each
GW Instek GPR-3510HD High-Current, Single-Output DC Power Supply, 0 to 35 VDC, 0 to 10 A WW-26856-94 Mfr # GPR-3510HD
COMPARE
0 to 10 A 0 to 35 VDC ≤ 0.5 mVrms; ≤ 3 mArms
₹188,000.00INR / Each