Instek GPD3303 Series DC Programmable Power Supplies
Minimum 1mV / 1mA resolution for highly accurate control of power sources
  • Three independent outputs - 30V/3A x 2, 2.5V/3.3V/5V/3A x 1 (fixed)
  • 4 LED displays with 3 digits after decimal point
  • Intuitive digital panel control with rotary encoder switch and rubber key with indicator
  • Coarse/fine volume control provides easy operation
  • 4 set save/recall function
  • Output on/off with audible warning buzzer
  • Tracking series and parallel mode
  • Smart cooling fan achieving low noise
  • Light-weight, compact design with key lock for additional flexibility
  • USB interface
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
GW Instek GPD-3303S Multi Output Programmable Linear DC Power Supply, 30V, 3A WW-20033-62 Mfr # GPD-3303S
COMPARE
0 to 3 A, 3 A 0 to 30.0 V, 2.5 VDC, 3.3 VDC, 5.0 VDC < 1 mVrms (5 Hz to 1 MHz)
₹154,000.00INR / Each