This product is now discontinued, View Related Products
Instek GDS-200 Series Compact Portable Digital Storage Oscilloscopes
Built-in digital multimeter works simultaneously with oscilloscope functions for a complete test and measurement instrument
  • Full touch panel, 7" (18 cm) LCD allows you to easily make waveform adjustments and set trigger conditions
  • Built-in engineering calculator and trend plot enable field analysis and debugging
  • Two input channels provide versatility
  • View results in either landscape or portrait orientation
The GDS-200 series feature a 5000-count digital multimeter and memory depth of one million points per channel.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Bandwidth (MHz) Channels Sample Rate (GS/s)
Availability Pricing
GW Instek GDS-207 Portable Digital Oscilloscope with 5,000 Count DMM, 70 MHz, 2 channels WW-20050-00 Mfr # GDS-207
COMPARE
70 2 1
Call For Price
GW Instek GDS-210 Portable Digital Oscilloscope with 5,000 Count DMM, 100 MHz, 2 channel WW-20050-01 Mfr # GDS-210
COMPARE
100 2 1
Call For Price
GW Instek GDS-220 Portable Digital Oscilloscope with 5,000 Count DMM, 200 MHz, 2 channel WW-20050-02 Mfr # GDS-220
COMPARE
200 2 1