This product is now discontinued, View Related Products
Instek Advanced Analog Oscilloscopes
Variable holdoff stabilizes the display of a composite signal
  • TV-sync function measures video signals
  • Trigger level controls
These oscilloscopes measure voltage, frequency, rise and fall time, and pulse width. Use the dual trace or the X-Y mode to measure the phase difference between two signals. Large 6'' CRT screen with 8 x 10 internal graticules provides distortion-free waveform measurements without parallax error.
Model 26857-14 features standard trigger modes including auto, norm, ext, and TV sync.
5 mV/div sensitivity extends the measurement range increasing test accuracies.
Model 26857-16 a 20 MHz bandwidth. Read up to two signals simultaneously—the oscilloscope features dual channel, dual trace, and X-Y mode.
Model 26857-30 has a 50 MHz bandwidth. This dual-channel scope features a delayed sweep function that lets you start a sweep at any point of the wave.
Model 26857-40 has a 100 MHz bandwidth. This oscilliscope provides real-time signal data in three dimensions — amplitude, time and distribution of amplitude over time.
Model 26857-50 has a 200 MHz bandwidth. Scope features dual channel and delayed sweep — start your sweep at any point in the wave.
What’s included: two probes (1x to 10x) and a 6-ft power cord with three-prong plug.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Bandwidth (MHz) Channels Sample Rate (GS/s)
Availability Pricing
GW Instek GOS-310 Model , Single Channel Oscilloscope, 10 MHz WW-26857-14 Mfr # GOS-310
COMPARE
10 1
Call For Price
GW Instek GOS-622G 20MHz Advanced Oscilloscope, 120/220 VAC WW-26857-16 Mfr # GOS-6620
COMPARE
20 2
Call For Price
GW Instek GOS-653G 50MHz Advanced Oscilloscope, 120/220 VAC WW-26857-30 Mfr # GOS-653G
COMPARE
50 2
Call For Price
GW Instek GOS-6103 Advanced Oscilloscope; 100 MHz Bandwidth WW-26857-40 Mfr # GOS-6103
COMPARE
100 1
Call For Price
GW Instek Advanced Oscilloscope; 200 MHz Bandwidth WW-26857-50 Mfr # GOS-6200
COMPARE
200