Idex® In-line Solvent Filter
In-line Solvent Filters for running cleaner systems
  • Help prevent particulates from clogging equipment
  • Parts made of stainless steel and PEEK®
  • Replacement frits available
In-line solvent filters are specifically engineered to help prevent particulate contamination from clogging sensitive equipment. Ideally, they should be placed along the flow path line between the pump and injection valve/autosampler. Available in a variety of porosities, filters threaded to fit 1/16" OD tubing, allowing the use of most standard chromatography high-pressure fittings.
4,700.00 - ₹19,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Pore Size (µm) Description
Availability Pricing
Idex Inline Solvent Filter Assembly, SS, 2 µm, PEEK Ring, 1/16" OD, 10-32 Coned; 1/EA WW-02010-00 Mfr # A-314
COMPARE
10-32 Coned 2 Inline Solvent Filter Assembly, SS, 2 µm, PEEK Ring, 1/16\" OD, 10-32 Coned; 1/EA
₹19,000.00INR / Each
Idex Inline Solvent Filter Assembly, SS, 20 µm, PEEK Ring, 1/16" OD; 1/EA WW-02010-03 Mfr # A-313
COMPARE
1/16\" OD 20 Inline Solvent Filter Assembly, SS, 20 µm, PEEK Ring, 1/16\" OD
₹14,600.00INR / Each
Idex Inline Solvent Filter, Female End, 2 µm, 1/16" OD; 1/EA WW-02010-04 Mfr # A-314-01
COMPARE
Female End 2 Inline Solvent Filter, Female End, 2 µm, 1/16\" OD; 1/EA
₹7,100.00INR / Each
Idex Inline Solvent Filter, Male End, 2 µm, 1/16" OD; 1/EA WW-02010-05 Mfr # A-314-02
COMPARE
Male end 2 Inline Solvent Filter, Male End, 2 µm, 1/16\" OD; 1/EA
₹5,900.00INR / Each
Idex Inline Solvent Filter, UHMWPE, 10 µm, 1/16" OD, 1/4-28 Flat Bottom; 1/EA WW-02010-07 Mfr # A-425
COMPARE
1/4-28 Flat Bottom 10 Inline Solvent Filter, UHMWPE, 10 µm, 1/16\" OD, 1/4-28 Flat Bottom; 1/EA
₹4,700.00INR / Each
Idex Inline Solvent Filter End Fitting, Black PEEK, 0.5 µm, 1/16" OD, 10-32 Coned; 10/PK WW-02010-08 Mfr # A-428X
COMPARE
10-32 Coned 0.5 Inline Solvent Filter End Fitting, Black PEEK, 0.5 µm, 1/16\" OD, 10-32 Coned; 10/PK
₹15,900.00INR / Pkg of 10
Idex Inline Solvent Filter End Fitting, Natural PEEK, 2 µm, 1/16" OD Tubing, 10-32 Coned; 10/PK WW-02010-09 Mfr # A-429X
COMPARE
10-32 Coned 2 Inline Solvent Filter End Fitting, Natural PEEK, 2 µm, 1/16\" OD Tubing, 10-32 Coned; 10/PK
₹16,300.00INR / Pkg of 10
Idex Biocompatible Inline Solvent Filter Assembly, PEEK/SS, 2 µm, 1/16" OD Tubing, 10-32 Coned; 1/EA WW-02010-10 Mfr # A-430
COMPARE
10-32 Coned 2 Biocompatible Inline Solvent Filter Assembly, PEEK/SS, 2 µm, 1/16\" OD Tubing, 10-32 Coned; 1/EA
₹7,900.00INR / Each
Idex Biocompatible Inline Solvent Filter Assembly, PEEK/SS, 0.5 µm, 1/16" OD Tubing, 10-32 Coned; 1/EA WW-02010-11 Mfr # A-431
COMPARE
10-32 Coned 0.5 Biocompatible Inline Solvent Filter Assembly, PEEK/SS, 0.5 µm, 1/16\" OD Tubing, 10-32 Coned; 1/EA
₹11,900.00INR / Each
Showing 1- 9 of 9