This product is now discontinued, View Related Products
IKA Trayster Overhead Shakers
Vertical rotation for smooth and effective mixing
These overhead rotating shakers are ideal for separating and mixing powdered, liquid, and biological samples in Eppendorf and Greiner tube sizes up to 50 mL. Basic and digital models are suitable for continuous operation. Shakers have a simple-to-use keypad and removable attachments for easy cleanup.
Trayster does not ship with Tube inserts, they must be purchased separately from selection below
79,766.00 - ₹87,924.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm)
Availability Pricing
IKA Trayster basic Trayster Overhead Shaker, Basic WW-04304-08 Mfr # 0004005000
COMPARE
20
₹79,766.00INR / Each
IKA 4006000 Trayster Overhead Shaker, Digital WW-04304-09 Mfr # 4006000
COMPARE
80
₹87,924.00INR / Each