IKA RW 28 Digital Pilot-Process Mixer
Powerful mechanically-controlled mixer for volumes up to 21 gallons (80 liters) and viscosities up to 50,000 cps
  • Two selectable speed ranges for high viscosity or high-speed mixing
  • Through-shaft design allows you to easily raise or lower the shaft and impeller
These mixers feature a through-shaft design that permits adjustment of shaft length and impeller depth—allowing you to place or remove mixing vessels without repositioning the mixer. Easy-to-use controls and a clear LED display of rpm enable you to precisely control and repeat your process parameters. Mixers feature infinitely adjustable stepless speed control.
Quiet, continuous-duty motors feature thermal overload shut-off and an efficient heat-dissipating design to ensure a long service life. Safety circuits ensure automatic cut-off in an anti-stall or overload condition.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
IKA 5040001 Digital Pilot-Process Mixer, Dual-Range, 115 VAC WW-50702-70 Mfr # 5040001
COMPARE
1400 1275 115