IKA RCT and RET basic IKAMAG Round Top Digital Stirring Hot Plates
Safety magnetic stirrers with heating — suitable for unsupervised operation
  • Temperature control mode for faster heating of medium
  • Platinum RTD temperature sensor ensures precise temperature control
Both units have a high safety level due to their improved heat control technology. Digitally set safety temperature limit. Hot top indicator warns to prevent burns.
RCT basic ™ features a polished aluminum round top plate which provides more efficient heat transfer than standard top plates. Hot plate features an over-temperature safety circuit adjustable from 122 to 680°F (50 to 360°C) to protect your samples from overheating. Adjustable speed up to 1500 rpm.
RET basic ™ models off the same features of the RCT models plus: a stainless steel top plate for better chemical resistance; a higher temerature range from 122-716°F (50 to 380°C) and a higher speed range — up to 1700 rpm for more demanding applications.
335,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Max Temperature (° F) Power (VAC)
Availability Pricing
IKA 3622001 RET basic MAG Stirring Hot Plate 115V WW-04671-25 Mfr # 3622001
COMPARE
1700 644 115
₹335,000.00INR / Each