This product is now discontinued, View Related Products
IKA® LR-2.ST Laboratory Reactors
Suitable for processing high-viscosity samples at high temperatures
  • Stand systems with EUROSTAR stirrers allow for controlled speed regulation
  • Different models include distinct EUROSTAR stirrers for various control styles
  • Ease of use is enhanced with removeable wireless controllers
These three setups allow for a composed user experience when reproducing, mixing, or homogenizing for a chemical reaction. The stations are constructed with a stand system to give a larger useable volume. Each item has solvent and temperature-resistant seals so contact with chemical samples will not degrade the machinery. All stations are appropriate for vacuum operation.
Each reaction station gives the user the option for torque trend display through the EUROSTAR stirrer to measure changing viscosity. The models each have a different EUROSTAR stirrer which allows the user to work with samples of different viscosity. The removable wireless controller is a feature of each setup and allows for safe and simple remote use.
5,122,000.00 - ₹5,692,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
IKA LR-2.ST IKA® Laboratory Reactor, High Performer; 115 VAC WW-28711-00 Mfr # 0020013404
COMPARE
230 115 High viscosity
₹5,692,000.00INR / Each
IKA LR-2.ST IKA® Laboratory Reactor, Allrounder; 115 VAC WW-28711-01 Mfr # 0020013402
COMPARE
230 115 Medium viscosity
₹5,285,000.00INR / Each
IKA LR-2.ST IKA® Laboratory Reactor, Versatile; 115 VAC WW-28711-02 Mfr # 0020013408
COMPARE
230 115 Low viscosity
₹5,122,000.00INR / Each