IKA Loopster Rotating Shakers
Adjust turntable from 0 to 90°
Rotating shakers with smooth and effective mixing are ideal for biological, powdered, and liquid samples in a variety of tube sizes up to 50 mL. Choose from basic or digital models. Both feature simple-to-use keypad control, removable turntable, and adjustable angle from 0 to 90°.
Loopster does not ship with a turntable Tube attachment, one must be purchased separately from selection below
88,523.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm)
Availability Pricing
IKA 4016000 Loopster Rotating Shaker, Digital WW-04304-13 Mfr # 0004016000
COMPARE
80
₹88,523.00INR / Each