This product is now discontinued, View Related Products
IKA C1 Calorimeters
Portable calorimeter is a little giant for isoperibol testing
  • Various interfaces allow connections to networks, PCs, balances, keyboards and more
  • User-friendly CalWin® C 6040 software available for controlling calorimeter and administration of measured data
This little giant in calorimeters is a portable isoperibol static jacket calorimeter featuring a high level of
automation with extremely small footprint. It features automatic ignition, automatic water filling and draining, automatic oxygen filling, venting and flushing; and computer interface using optional CalWin C 6040 software (order separately) for controlling the calorimeter and administration of
measuring data. The C 1 calorimeter will interface with a PC via USB, or printer/balance via a serial interface.
The traditionally heavy screw-threaded bomb has been replaced by a light attachable combustion chamber. To provide the calorimeter with cooling water, it needs to be connected to a
thermostat (order models 50906-00 or -02 with chiller) or a firmly installed water connection.
Models 50906-02 and -06 include a special halogen-resistant vessel, in lieu of the standard vessel, for quantitative decomposition of halogens and sulfur. The crucible holders can be changed over to use combustible crucibles, which are sold separately. Models 50906-04 and -06 are sold with a water heater, which can run with standard tap water.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Max Temperature (° C) Power (VAC)
Availability Pricing
IKA 8804901 C 1, S 1 Package 1/10 Calorimeter WW-50906-00 Mfr # 0010001059
COMPARE
Call For Price
IKA 8805001 C 1, S 1 halogen resistant Package 1/12 Calorimeter WW-50906-02 Mfr # 8805001
COMPARE
Call For Price
IKA 8805101 C 1, S 1 Package 2/10 Calorimeter WW-50906-04 Mfr # 8805100
COMPARE
Call For Price
IKA 8805201 C 1, S 1 halogen resistant Package 2/12 Calorimeter WW-50906-06 Mfr # 8805201