This product is now discontinued, View Related Products
Idex® Stainless Steel Sample Loops for Injection Valve Models 8125 and 8126
Square-cut, burr-free ends provide a firm, flush connection to valve ports
  • 316 stainless steel construction
  • High pressure resistance, low permeability
A sample loop is a defined length of tubing and fittings configured to match the angle of the injection valve ports. A sample is loaded and held in the loop until injection is triggered, either automatically or manually. Stainless steel loops are supplied with unswaged fittings, so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal.
17,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (µL) Tubing Inside Diameter (ID) (mm) Tubing Outside Diameter (OD) (in)
Availability Pricing
Idex Sample Loop for 8125/8126 Valves, Stainless Steel, 5 µL, 0.020" OD x 0.008" ID WW-02016-43 Mfr # 8020
COMPARE
5 0.2 0.02
Call For Price
Idex Sample Loop for 8125/8126 Valves, SS, 10 µL, 0.020" OD x 0.008" ID; 1/EA WW-02016-44 Mfr # 8021
COMPARE
10 0.2 0.02
₹17,300.00INR / Each
Idex 8022 Sample Loop for 8125/8126 Valves, SS, 20 µL, 0.020" OD x 0.010" ID; 1/EA WW-02016-45 Mfr # 8022
COMPARE
20 0.25 0.02
Call For Price
Idex 8023 Sample Loop for 8125/8126 Valves, SS, 50 µL, 0.020" OD x 0.012" ID; 1/EA WW-02016-46 Mfr # 8023
COMPARE
50 0.3 0.02