Idex® Manual Chromatography Selection Valves
Allow operator control of low-use application valves
  • Enable connections among multiple fluid systems 6-position, 7-port configuration
Manual valves serve well in applications that require operator control or involve lesser frequency of use. Selection valves enable connection to one column for another without the need to manually disconnect the plumbing. In sequencing or diagnostic applications, the valve alternates between the sample or reagent streams.
166,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description Tubing OD (in) Wetted Materials
Availability Pricing
Idex Manual Selection Valve, Analytical, SS/Vespel®, 6-Position, 7-Port, 1/16" OD Tubing; 1/EA WW-02015-24 Mfr # 7060
COMPARE
Manual Selection Valve, 6-Way, SS/Vespel®, 6-Position, 7-Port, 1/16\" OD Tubing; 1/EA 0.0625 SS/Vespel
₹166,000.00INR / Each