This product is now discontinued, View Related Products

IDEC DIN-Rail Power Supplies

Compact design provides exceptional power output for all system needs
  • Save valuable DIN-rail space while filling your power requirements
These switching power supplies offer the versatility of DIN-rail or direct surface mounting. The unique spring-up, terminals feature an IP20 (finger-safe) rating — ideal for ring lug terminated wires. Adjust output voltage with easy access trim pot to +/-10%. Outputs also feature auto-resetting overcurrent protection and 2% regulated maximum ripple. Other features include UL508 listing (no derating), fused inputs and an LED verifying power.
Models 26901-08, -10, -12, and –16 provide UL1604 listing for hazardous locations and Class 2 rating (-08, -10, and –12). Models 26901-14, -18, and –20 offer the capability of being connected in parallel to increase the current output.
Call For Price

11 variations of this product are available.

See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
Idec PS5R-A12 DIN-Rail Power Supply 7.5W (12 VDC, 0.6 A) WW-26901-00 Mfr # PS5R-A12
COMPARE
0.6 A 12 VDC ≤ 2%
Call For Price
Idec PS5R-A24 DIN-Rail Power Supply 7.5W (24 VDC, 0.3 A) WW-26901-02 Mfr # PS5R-A24
COMPARE
0.3 A 24 VDC ≤ 2%
Call For Price
Idec PS5R-B12 DIN-Rail Power Supply 15W (12 VDC, 1.2 A) WW-26901-04 Mfr # PS5R-B12
COMPARE
1.2 A 12 VDC ≤ 2%
Call For Price
Idec PS5R-B24 DIN-Rail Power Supply 15W (24 VDC, 0.6 A) WW-26901-06 Mfr # PS5R-VB24
COMPARE
0.6 A 24 VDC ≤ 2%
Call For Price
Idec PS5R-SC12 DIN-Rail Power Supply 30W (12 VDC, 2.5 A) WW-26901-08 Mfr # PS5R-SC12
COMPARE
2.5 A 12 VDC ≤ 2%
Call For Price
Idec PS5R-SC24 DIN-Rail Power Supply 30W (24 VDC, 1.3 A) WW-26901-10 Mfr # PS5R-VC24
COMPARE
1.3 A 24 VDC ≤ 2%
Call For Price
Idec PS5R-SD24 DIN-Rail Power Supply 60W (24 VDC, 2.5 A) WW-26901-12 Mfr # PS5R-SD24
COMPARE
2.5 A 24 VDC ≤ 2%
Call For Price
Idec PS5R-Q24 DIN-Rail Power Supply 75W (24 VDC, 3.1 A) WW-26901-14 Mfr # PS5R-Q24
COMPARE
3.1 A 24 VDC ≤ 2%
Call For Price
Idec PS5R-SE24 DIN-Rail Power Supply 90W (24 VDC, 3.75 A) WW-26901-16 Mfr # PS5R-SE24
COMPARE
3.75 A 24 VDC ≤ 2%
Call For Price
Idec PS5R-F24 DIN-Rail Power Supply 120W (24 VDC, 5 A) WW-26901-18 Mfr # PS5R-F24
COMPARE
5 A 24 VDC ≤ 2%
Call For Price
Idec PS5R-G24 DIN-Rail Power Supply 240W (24 VDC, 10 A) WW-26901-20 Mfr # PS5R-G24
COMPARE
10 A 24 VDC ≤ 2.0%
Call For Price
Showing 1- 11 of 11

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat