Wildco® Horizontal Alpha Water Samplers
Suitable for use in oceans, deep lakes, and corrosive waters
  • Urethane end seals snap shut with minimum surface disturbance on messenger contact Drain valve provides easy sample removal
Choose horizontal samplers for sampling at the surface, thermocline, or just above the bottom. Samplers include a carrying case; order messenger and line separately. Silicone end seals are available by special order.
141,000.00 - ₹163,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Material
Availability Pricing
Wildco 1120-H40 Horizontal alpha water sampler, PVC, 2.2 L WW-05488-00 Mfr # 1120-H40
COMPARE
Horizontal alpha water sampler, PVC, 2.2 L PVC
₹141,000.00INR / Each
Wildco 1120-G40 Horizontal alpha water sampler, acrylic, 2.2 L WW-05488-10 Mfr # 1120-G40
COMPARE
Horizontal alpha water sampler, acrylic, 2.2 L Acrylic
₹163,000.00INR / Each