Cole-Parmer® Essentials Hinged-Lid Sample Containers, PP
Hinged cap design allows you to easily close lid with one finger
  • FDA compliant
  • Autoclavable
Made from polypropylene (PP), these containers are ideal for storing liquids or solids and for metals analysis. When containers are properly closed, your sample won't spill or leak in any position, reducing the chance of contamination.
71,900.00 - ₹181,000.00INR / Pkg of 2500
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (oz) Material Overall Height (in)
Availability Pricing
COMPARE
0.25 Polypropylene 5/8
₹171,000.00INR / Pkg of 2500
COMPARE
0.5 Polypropylene 3/4
₹181,000.00INR / Pkg of 2000
COMPARE
0.75 Polypropylene 1
₹160,000.00INR / Pkg of 1500
COMPARE
1 Polypropylene 1 1/2
₹127,000.00INR / Pkg of 1300
COMPARE
2 Polypropylene 1 5/8
₹111,000.00INR / Pkg of 750
COMPARE
4 Polypropylene 3 3/16
₹71,900.00INR / Pkg of 400