These Ashcroft Series GC35 digital pressure sensors combine a digital gauge, switch, and transducer in one package. Simple push-button menu allows you to configure switch settings and analog scaling. Unique high-visibility LED display surrounds the sensor to alert you to switch status. A green LED signals pressure is within pressure range, while a red LED alerts you to pressure out-of-switch range. Configure the analog output scaling to any range within full-scale of sensor range, and the switch setting can be adjusted to any actuation and deadband setting within full-scale sensor range via keypad. You can also use the keypad to reconfigure switch contacts to either NPN or PNP, display scaling, set to capture min/max value, and adjust display update rate.
Applications Hydraulic presses, stamping equipment, lifts Tire press vulcanization pressure monitoring Water, refrigerant, or ammonia based cooling systems Pressure monitoring on lubrication systems