High-Vacuum Pumps for Corrosive Gases
  • Corrosion-resistant parts—ideal for pumping corrosive gases such as Cl2, HBr, F3CCO2H, and HOAC
  • Pumps draw clean oil from top of oil reservoir, not from the bottom as do direct-drive pumps
  • Large oil reservoir dilutes contaminants
  • Lower pump speed (less than 580 rpm) results in less wear, cooler operation, and longer service life
  • Bubbler tube in the oil reservoir allows nitrogen gas purging, resulting in lower operating temperature, reduced corrosion rate, and extended pump oil life
Pumps include a clamp, centering ring, NW flange to hose barb fitting, pump oil, and an instruction manual containing an applications booklet.
560,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Power (VAC)
Availability Pricing
Welch 1400N-50 Rotary Vane Vacuum Pump for Corrosive Gases, 0.9 cfm, 230 VAC WW-79201-05 Mfr # 1400N-50
COMPARE
0.9 230
₹560,000.00INR / Each
Welch 1402N-50 Rotary Vane Vacuum Pump for Corrosive Gases, 5.6 cfm, 230 VAC WW-79201-15 Mfr # 1402N-50
COMPARE
5.6 230