High-Tech Conversions Dry Sterile Cleanroom Wipes
Gamma irradiated to a 10-6 sterility assurance level
  • Cleanroom ISO class 5 compliant Lot numbered for traceability Meets USP 797 and 800 requirements for critical contamination control
The Cobalt Blue dry sterile cleanroom wipes are comprised of a virgin polyester and cellulose blend, offering durability, absorbency, and abrasion resistance. Wipes clean without compromising the sterile nature of the controlled environment. All wipes are gamma irradiated with Cobalt 60 and validated to Sterility Assurance Level (SAL) of 10-6 in accordance with standards and recommended practices of the ANSI/AAMI/ISO 11137:2006 guidelines. Each bag of wipes is identified with a lot number for traceability and a Certificate of Sterility is included with every case.
83,700.00 - ₹84,700.00INR / Case of 1500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compliance Material Width (in)
Availability Pricing
High-Tech Conversions Cleanroom wipes, sterile, dry, Non-woven poly-cellulose, 9" x 9", 1500/CS WW-15981-04 Mfr # NT10-99CB
COMPARE
ISO Class 5 Polyester/Cellulose 9
₹84,700.00INR / Case of 1500
High-Tech Conversions Cleanroom wipes, sterile, dry, Non-woven poly-cellulose, 12" x 12", 840/CS WW-15981-05 Mfr # NT10-1212CB
COMPARE
ISO Class 5 Polyester/Cellulose 12
₹83,700.00INR / Case of 840