High-Tech Conversions Disposable Cleaning Pads
Highly absorbent – suitable for cleanroom applications
  • No chemical dyes used in manufacturing process
  • Single use application reduces risk of cross contamination
  • Compatible with chlorine bleach, hydrogen peroxide, and common disinfectants
  • Channels efficiently pick up and retain dirt in pad
  • Bonded edges improve performance, reduces linting
  • Bag in bag packaging for cleanroom application
  • No quat binding
These Virtu-Clean™ disposable cleaning pads are designed for critical environments, giving you maximum cleaning coverage. The single-use mop pads hold more liquid and release that liquid at a controlled rate over a large surface area. The embossed pattern on the pads reduces friction providing you a more effort-free cleaning.
37,300.00 - ₹41,300.00INR / Case of 240
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compliance Length (in) Description
Availability Pricing
High-Tech Conversions Virtu-Clean® Disposable Cleaning Pads; 20/Bag, 12 Bags/CS WW-09411-95 Mfr # VCDC
COMPARE
ISO Class 6, USP 797 compliant 19 Virtu-Clean® Disposable Cleaning Pads; 20/Bag, 12 Bags/CS
₹37,300.00INR / Case of 240
High-Tech Conversions Virtu-Clean® Disposable Cleaning Pads with Scrubber; 20/Bag, 12 Bags/CS WW-09411-96 Mfr # VCDCS
COMPARE
ISO Class 5, USP 797 compliant 19 Virtu-Clean® Disposable Cleaning Pads with Scrubber; 20/Bag, 12 Bags/CS
₹41,300.00INR / Case of 240