Cole-Parmer® Single-Stage Gas Regulators
Ideal for corrosive or noncorrosive gas applications. Designed and constructed for diffusion resistance and easy cleanup. Only brass, stainless steel (SS), PTFE, and Kel-F� contact the operating media.
Metal diaphragm, packless valves, and high-purity seals minimize or eliminate outgoing and inboard diffusion. Supplied with SS screen filter (25 µm); and brass body, bonnet, and gauges. Single-Stage High-Purity Regulators are recommended for analytical work where precise pressure control is not critical.
122,000.00 - ₹126,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (SCFH) Max Delivery Pressure (psig) Max Inlet Pressure (PSI)
Availability Pricing
COMPARE
500 50 3000
₹122,000.00INR / Each
COMPARE
500 50 3000
₹122,000.00INR / Each
COMPARE
500 50 3000
₹122,000.00INR / Each
COMPARE
1000 100 3000
₹126,000.00INR / Each
COMPARE
1000 100 3000
₹126,000.00INR / Each