Nederman High-Efficiency Fume Extractor Arms
Up to 650 cfm air capacity for particles, smoke, and dust
Fume extractor arms are perfect for welding booths and other industrial applications requiring large air capacities. Arms come in four lengths: 7 ft; 10 ft; 14 ft; and telescopic, which measures 34" when retracted, 5 ft when fully extended. Telescopic and 7-ft arms are suitable for small work areas; 10-ft and 14-ft arms are best for wider work areas. Specially designed friction joints allow hoods to be positioned at any point within the work area. Arms are supported by a 360deg swivel elbow for unlimited maneuverability. Maximum operating temperature is 158degF (70degC). Assembly is required for all models.
The 7-ft, 10-ft, and 14-ft arms include 10" x 12" oval polycarbonate hood with handle and damper, and 6-1/4" OD PVC fiberglass-reinforced hose. Telescopic a
32,700.00 - ₹607,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Exhaust Volume (CFM) Description
Availability Pricing
Nederman High-Efficiency Fume Extractor Arm, 84"L WW-33700-01 Mfr # 10554235
COMPARE
650 High-Efficiency Fume Extractor Arm, 84\" L
₹314,000.00INR / Each
Nederman High-Efficiency Fume Extractor Arm, 120"L WW-33700-02 Mfr # 10554335
COMPARE
650 High-Efficiency Fume Extractor Arm, 120\" L
₹364,000.00INR / Each
Nederman High-Efficiency Fume Extractor Arm, 168"L WW-33700-04 Mfr # 10554535
COMPARE
650 High-Efficiency Fume Extractor Arm, 168\" L
₹397,000.00INR / Each
Nederman High-Efficiency Telescopic Fume Extractor Arm, 32 to 60"L WW-33700-08 Mfr # 10502531
COMPARE
650 High-Efficiency Telescopic Fume Extractor Arm, 32 to 60\" L
₹285,000.00INR / Each
Nederman Extractor Arm, Telescopic, 32" to 5ft, 6" diameter WW-33700-09 Mfr # 10502731
COMPARE
650 Extractor Arm, Telescopic, 32\" to 66\" diameter
₹431,000.00INR / Each
Nederman High-Efficiency Fume Extractor Arm with Light, 84"L WW-33700-10 Mfr # 1055-4235,-1235,14513637
COMPARE
650 High-Efficiency Fume Extractor Arm with Light, 84\" L
₹607,000.00INR / Each
Nederman High-Efficiency Fume Extractor Arm with Light, 168"L WW-33700-14 Mfr # 1055-4535,10376922,14518937
COMPARE
650 High-Efficiency Fume Extractor Arm with Light, 168\" L
₹594,000.00INR / Each
Nederman Fan for Fume Extractor Arms, 3/4 hp, 115/230 VAC WW-33700-50 Mfr # 14510321
COMPARE
300 to 400 Fan for Fume Extractor Arms, 3/4 hp, 115/230 VAC
₹292,000.00INR / Each
Nederman Fan for Fume Extractor Arms, 1 hp, 115/230 VAC WW-33700-60 Mfr # 14510222
COMPARE
550 to 650 Fan for Fume Extractor Arms, 1 hp, 115/230 VAC
₹501,000.00INR / Each
Nederman Wall Bracket for 33700 Series Extractor Arms WW-33700-90 Mfr # 10550635
COMPARE
Wall Bracket for 33700 Series Extractor Arms
₹32,700.00INR / Each
Showing 1- 10 of 10