Scienceware® High-Capacity Polypropylene Beakers
Easy pouring of large samples
  • Easy dispensing with flared rim and pouring spout
  • Autoclavable
The largest plastic beakers we sell! Choose 5- or 10-liter high-capacity polypropylene (PP) beakers. These giant-sized beakers feature flared rims and pouring spouts for quick, easy dispensing. Beakers offer good chemical resistance and withstand temperatures to 250°F (121°C). They are not graduated.
20,500.00 - ₹37,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material
Availability Pricing
Scienceware 26219-0005 high-capacity polypropylene beaker, 5 liter WW-06147-00 Mfr # 26219-0005
COMPARE
5 Polypropylene(PP)
₹20,500.00INR / Each
Scienceware 26219-0010 high-capacity polypropylene beaker, 10 liter WW-06147-10 Mfr # 26219-0010
COMPARE
10 Polypropylene(PP)
₹37,600.00INR / Each