This product is now discontinued, View Related Products
High-Absorption Desiccants
High-Absorption Desiccant/Purifier
  • Indicator desiccant absorbs 107% of its weight in water
  • 4 to 8 mesh
This indicating desiccant (alumina silicate) quickly absorbs 107% of its own weight in water and dries to a dew point of –100°F. It reduces moisture in air or gas to 17% in a room with a temperature of 70°F and relative humidity of 40%. Desiccant can be repeatedly reactivated in a furnace, dehumidifier, or microwave oven. In addition to drying air, desiccant removes harmful substances from air or liquids. Pore sizes range from 2.6 to 5.0 Å.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
High-absorption desiccant/purifier, 1 lb bottle WW-24450-00 Mfr # MG1-6
COMPARE
High-absorption desiccant/purifier, 1 lb bottle
Call For Price
COMPARE
High-absorption desiccant/purifier, 5 lb bag
Call For Price
High-absorption desiccant/purifier, 55 lb bag WW-24450-20 Mfr # MG-BULK
COMPARE
High-absorption desiccant/purifier, 55 lb bag