This product is now discontinued, View Related Products
HF Scientific MicroTOL On-Line Turbidimeters
Specially designed system eliminates the need for a bubble trap and ensures immediate response time
  • Optical design increases accuracy while providing more consistent readings
  • Low, 30-mL sample volume allows for fast response time and inexpensive calibration
  • Two light sources available — white light to meet U.S. EPA method 180.1 regulations and infrared for ISO 7027
MicroTOL 2 Turbidimeters are ideal for measuring turbidity continuously in filtered water, raw water, waste water final effluent, and industrial applications. They arrive calibrated and verify readings in seconds. Versatile design allows for one-piece mounting for added ease of installation or multi-piece mounting for installation flexibility. Removable sample cuvettes allow for fast, easy cleaning and calibration. Optics do not contact the sample, reducing the chance of false, low readings.
MicroTOL 3 and 4 Turbidimeters build on the features of the MicroTOL 2. These units add the auto-clean ultrasonic cleaning system, which increases the interval between cleanings for finished or raw water applications.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Control Type Output Current
Availability Pricing
HF Scientific 20053 MicroTOL 2 Turbidimeter, 0 to 1000 NTU, EPA compliant WW-99544-00 Mfr # 20053
COMPARE
On/Off 4-20mA
Call For Price
HF Scientific 20054 MicroTOL 2 Turbidimeter, 0 to 1000 NTU, ISO compliant WW-99544-05 Mfr # 20054
COMPARE
On/Off 4-20mA
Call For Price
HF Scientific 20055 MicroTOL 3 Turbidimeter; 0 to 100 NTU/EPA/Self-Cleaning WW-99544-10 Mfr # 20055
COMPARE
On/Off 4-20mA
Call For Price
HF Scientific 20056 MicroTOL 3 Turbidimeter; 0 to 100 NTU/ISO/Self-Cleaning WW-99544-15 Mfr # 20056
COMPARE
On/Off 4-20mA
Call For Price
HF Scientific 20063 MicroTOL 4 Turbidimeter; 0 to 1000 NTU/EPA/Self-Cleaning WW-99544-20 Mfr # 20063
COMPARE
On/Off 4-20mA
Call For Price
HF Scientific 20064 MicroTOL 4 Turbidimeter; 0 to 1000 NTU/ISO/Self-Cleaning WW-99544-25 Mfr # 20064
COMPARE
On/Off 4-20mA