This product is now discontinued, View Related Products
HF Scientific Micro 100 Benchtop Turbidity Meters
Auto calibration alert ensures that your meter is calibrated for accurate measurements and simple calibration
  • Quick connection lamp module for low maintenance
These turbidity meters provide accuracy and reliability. With a resolution of 0.01 nephelometric turbidity units at low turbidity readings and an extended range to 1000 NTU, these benchtop turbidity meters are an ideal and affordable tool for research, as well as routine analytical measurements. RS-232 output downloads date, time, and NTU reading—can be captured with a serial printer or data recorder.
Model with infrared light source meets international standard ISO 7027 for turbidity measurement.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Turbidity Min Turbidity Turbidity Resolution
Availability Pricing
HF Scientific 20001 Micro 100 Benchtop Turbidity Meter, EPA compliant, 110 VAC WW-99543-00 Mfr # 20001
COMPARE
1000 0.01 0.01, 0.1, or 1 NTU
Call For Price
HF Scientific 19950 Micro 100 Benchtop Turbidity Meter, EPA compliant, 220 VAC WW-99543-05 Mfr # 19950
COMPARE
1000 0.01 0.01, 0.1, or 1 NTU
Call For Price
HF Scientific 19952 Micro 100 Benchtop Turbidity Meter, ISO compliant, 220 VAC WW-99543-15 Mfr # 19952
COMPARE
1000 0.01 0.01, 0.1, or 1 NTU