This product is now discontinued, View Related Products
HEPA Capsule and Venting Filters
Ideal for pharmaceutical, biotechnology, and food and beverage applications.
  • 99.97% efficiency at 0.3 µm
Designed to provide bacteria-, algae-, fungal-free air for sterile applications, such as fermentors or incubators. The media allows for high flow rates and low pressure drop. Designed for high retention and particle holding capacity for air and gas. Housing is made of durable polypropylene.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Effective Filtration Area (cm²)
Availability Pricing
ZenPure Hepa venting filter disc, 50 mm diameter, 10/pk WW-49700-90 Mfr # D50CG0052H2H-10
COMPARE
15.9
Call For Price
ZenPure Hepa capsule filter, 625 cm2 filtration area WW-49700-92 Mfr # SKLPHH2H2H
COMPARE
600
Call For Price
ZenPure Hepa capsule filter, 1200 cm2 filtration area WW-49700-94 Mfr # SKLPHS3H3H
COMPARE
1200