Neptune Heavy-Duty, Gear-Drive Explosion-Proof Electrically-Powered Mixers
Powerful mixers for difficult applications
  • Grease-filled gearbox eliminates oil seal and the possibility of oil contamination
  • Mixers have no clutches to slip, wear, or replace
  • Shaft bearings are permanently lubricated
Indexed ball-and-socket design with vibration pad lets you adjust the mixer position and achieve various process results you want. Cast aluminum housing.
Note: Does not include power cord, as it should be wired at customer's site by qualified technician in accordance with local electrical codes. Wiring diagram is displayed on motor. Models of 3⁄4- to 1-hp require a separate motor starter, please call our Application Specialists for details.
657,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Impeller Size (in) Max Torque (in-oz) Description
Availability Pricing
Neptune Heavy duty, gear-drive explosion proof mixer; 1/3-hp, 60"L shaft, 10" impeller WW-50311-40 Mfr # JG-2.2-60
COMPARE
10 1080 Heavy duty, gear-drive explosion proof mixer; 1/3-hp, 60\"L shaft, 10\" impeller
₹657,000.00INR / Each