Cole-Parmer Essentials Heat-Resistant Cable Zip Ties, Polypropylene
Heat- and chemical-resistant for challenging environments
  • Withstand temperatures to 115 °C (239 °F)
  • Resistant to chemicals and salts
  • Locking mechanism holds securely
  • Bent tail for easy pick up and use
Polypropylene cable zip ties provide excellent resistance to chemicals and salts. They also have a wide operating temperature of -40 °C to 115 °C (-40 °F to 239 °F) making them ideal in situations where high heat could lead to melting of a nylon cable tie. A bent tail allows for easy pick up of the tie and threading through the lock. These ties are RoHS compliant, UL 94 HB flammability rated, and autoclavable.
Easily install by wrapping the tie around the bundle and threading the tail through the locking mechanism. Pull tight by hand and cut the tail to complete or use a tensioning tool—available separately.
1,900.00 - ₹55,000.00INR / Pkg of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Tensile Strength (lbs)
Availability Pricing
COMPARE
White 4 8
₹40,000.00INR / Pkg of 1000
COMPARE
Black 8 25
₹55,000.00INR / Pkg of 1000
COMPARE
White 4 8
₹1,900.00INR / Pkg of 100
COMPARE
Black 8 25
₹1,900.00INR / Pkg of 100